VOL-C-RTD-2.1 for VOLS-1 is especially suitable for expensive and valuable samples. A temperature sensor is already integrated in the chamber and thus allows for temperature measurement during viscosity determination, required to use the VOL-SP-2.1 spindle in combination with the 2.1 ml chamber
# de catalogue 76499-088
Fournisseur: IKA Works
Customers Who Bought This Also Bought