VOL-SP-3.8 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 3.8 ml, It is required to use the 3.8 ml chamber VOL-C-RTD-3.8 in combination with the VOL-SP-3.8 spindle
# de catalogue 76499-084
Fournisseur: IKA Works
Customers Who Bought This Also Bought