ContactUsLinkComponent
VOL-SP-13.5 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 13.5 ml.
# de catalogue 76499-080
Fournisseur:  IKA Works
VOL-SP-13.5 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 13.5 ml.
# de catalogue 76499-080
Fournisseur:  IKA Works
Customers Who Bought This Also Bought