ContactUsLinkComponent
VOL-SP-16.1 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 16.1 ml.
# de catalogue 76499-072
Fournisseur:  IKA Works
VOL-SP-16.1 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 16.1 ml.
# de catalogue 76499-072
Fournisseur:  IKA Works
Customers Who Bought This Also Bought