VOL-SP-9.4 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 9.4 ml.
# de catalogue 76499-070
Fournisseur: IKA Works
VOL-SP-9.4 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 9.4 ml.
# de catalogue 76499-070
Fournisseur: IKA Works
Customers Who Bought This Also Bought
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
73,48 $ to 4 408,00 $
Masterflex® L/S® Precision Variable-Speed Console Drives, Avantor®
Fournisseur: Avantor Fluid Handling
3 286,40 $
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Fournisseur: VWR International
147,27 $ to 464,44 $