ContactUsLinkComponent
VOL-SP-9 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 9 ml.
# de catalogue 76499-068
Fournisseur:  IKA Works
VOL-SP-9 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 9 ml.
# de catalogue 76499-068
Fournisseur:  IKA Works
Customers Who Bought This Also Bought