SP-5, standard spindle, is suitable for measurement in an extended viscosity range. It is intended to be used in combination with the rotary viscometers ROTAVISC me-vi, hi-vi I or hi-vi II Complete or ROTAVISC me-vi, hi-vi I or hi-vi II Advanced.
# de catalogue 76499-064
Fournisseur: IKA Works
Customers Who Bought This Also Bought