Sp Set-1 Stand Spindle 22, Includes Four Spindles (Sp1, Sp2, Sp3, Sp4). It Is Particularly Suitable For Viscosity Determination In Combination With The Rotary Viscometers Rotavisc Lo-Vi Complete Or Rotavisc Lo-Vi Advanced
# de catalogue 76499-060
Fournisseur: IKA Works
Customers Who Bought This Also Bought