Green Fluorescent Protein Polyclonal antibody, Host: Chicken, Reactivity: Variants of recombinant Aequorea green-fluorescent proteins, Conjugate: Biotin, Material is lyophilized, Should be kept at 4 deg C until rehydrated and will remain stable for two years, Use:Western Blot/ELISA, Size: 100ug
# de catalogue 76499-946
Fournisseur: Columbia Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.