Igy (H+L) Polyclonal antibody, Host: Goat, Reactivity: Chicken, Conjugate: Biotin, Product is a liquid, Store at 2-8 deg C, Buffer: PBS, ph 7.2, Preservative: 0.09% sodium azide, is stable for two years, Use:Western Blot/ELISA, Size: 1Mg
# de catalogue 76499-944
Fournisseur: Columbia Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.