Gloves, 18 Gauge Blue Nylon/Carbon/Spandex Seamless Knit Liner, Blue Nitrile Rubber Palm Coat, Silicone & Latex Free, ESD Certified, TouchScreen Compatible, Long Lasting Fortix Grip Protection, Barehand Tactility, Sz 6
# de catalogue 76498-972
Fournisseur: Ansell Healthcare
Customers Who Bought This Also Bought