ContactUsLinkComponent
Gloves, Black Intercept (Kevlar, stainless steel, polyamide, spandex ), 15G Circular Knit (Patented Technology), Elasticized Top, Knitwrist Cuff, Narrow Width, Integrated elbow pad to protect against impact, Fiber glass free, Size: 18in
# de catalogue 76498-956
Fournisseur:  Ansell Healthcare
Gloves, Black Intercept (Kevlar, stainless steel, polyamide, spandex ), 15G Circular Knit (Patented Technology), Elasticized Top, Knitwrist Cuff, Narrow Width, Integrated elbow pad to protect against impact, Fiber glass free, Size: 18in
# de catalogue 76498-956
Fournisseur:  Ansell Healthcare
Customers Who Bought This Also Bought