Capmats for J.T.Baker® Plate Round Well, Ritter, part of Avantor
# de catalogue 43001-0017
Fournisseur: Ritter, part of Avantor
Spécifications
- Optical property:Transparent
- Sterility:Non sterile
- Description:Capmat for J.T.Baker® plate round well
- Environmentally Preferable:Low Manufacturing Impact
- Material:EVA
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Packaging:1 bag of 20
Spécifications
A propos de cet article
Suitable to use with J.T.Baker® Plate Round Well 43001-0010, 43001-0016 or 43001-0116.
Customers Who Bought This Also Bought