ContactUsLinkComponent
Capmat for plate 1 ml, shore 85
Capmats for J.T.Baker® Plate Round Well, Ritter, part of Avantor
# de catalogue 43001-0017
   Low Manufacturing Impact
Capmat for plate 1 ml, shore 85
Capmats for J.T.Baker® Plate Round Well, Ritter, part of Avantor
# de catalogue 43001-0017
   Low Manufacturing Impact
Spécifications
  • Optical property:
    Transparent
  • Sterility:
    Non sterile
  • Description:
    Capmat for J.T.Baker® plate round well
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Material:
    EVA
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Packaging:
    1 bag of 20
Spécifications
A propos de cet article
Suitable to use with J.T.Baker® Plate Round Well 43001-0010, 43001-0016 or 43001-0116.
Customers Who Bought This Also Bought