Spécifications
- Dimensions:110H×16Ø mm
- Description:Staining jar with plastic lid
- Cat. no.:76498-154
Spécifications
A propos de cet article
Heavy, clear glass jar with four molded internal grooves to hold up to ten slides.
- Glass base construction
- Four molded grooves in glass hold up to ten 3×1" slides
Jar features a broad base to prevent spillage. Both glass and plastic lid options protect against contaminants. The plastic option is a tight-fitting screw lid to prevent fluid evaporation. The glass cover option simply and easily sets upon the jar.