Sas-1, Spiral adapter to measure the viscosity of very sticky and other viscous samples. Additionally, it is possible to analyse the fluid behavior of different shear rates.
# de catalogue 76499-050
Fournisseur: IKA Works
Customers Who Bought This Also Bought