T-spindle set is particularly suitable for the measurement of non-flowing samples, such as creams, pastes or gelatin. The set is the right accessory for electrica HELISTAND.
# de catalogue 76499-026
Fournisseur: IKA Works
Customers Who Bought This Also Bought