Helistand, motorised, allows you to measure the viscosity of highly viscous samples with a high level of reliability. It moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample.
# de catalogue 76499-024
Fournisseur: IKA Works
Customers Who Bought This Also Bought