To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Handheld meter MU 6100 H S1 kit, with pH electrode and conductivity probe
- Model:MU 6100 H
- Temperature accuracy:±0.2 °C
- Temperature resolution:0.1 °C
- Temperature range:Auto: −5.0...+105.0; Manual: −25...+130 °C
- Automatic buffer recognition:BDH Prolabo® TITRINORM® buffers at 25 °C: 4.00/6.98/9.94
NIST/DIN 19266 (1.68/4.00/6.86/9.18/12.54)
Technical buffers at 25 °C: 4.00/7.00/10.00 - Compliance:IP 67/GLP
- Conductivity accuracy:±0.5% of measured value
- Conductivity cell constant:Depending on probe
- Conductivity resolution:0.1 µS/cm
- Connections:BNC for pH/banana for temperature/8-pin for conductivity and DO
- DO calibration:Water saturated air
- DO concentration:0.00 - 20.00 / 0.0 - 90.0 mg/L
- DO concentration resolution:0.1%; 0.01 mg/L
- DO saturation:0.0 - 200.0%; 0 - 600%
- DO accuracy:±0.5% of value (between 5...30 °C)
- Data logging capacity:5000 data sets
- Data logging - log function:Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
- Display:LCD graphic display with LED backlight
- Power supply:4× AA batteries
- Resistivity range:1 Ω/cm - 999 MΩ/cm
- Resistivity accuracy:±0.5% of measured value
- Salinity range:0.00 - 70 ppt
- TDS range:0 - 1999 mg/L / 2.00 - 19.99 g/L / 20.0 - 199.9 g/L
- TDS resolution:1 mg/L / 0.01 g/L / 0.1 g/L
- Temperature compensation:Automatic/manual
- mV accuracy:±0.3/±1
- mV range:±1200.0/±2500
- mV resolution:0.1/1
- pH accuracy:±0.005 + 1 digit
- pH calibration:Up to 5 points
- pH range:−2.000...+20.000
- pH resolution:0.1/0.01/0.001 (selectable)
- Cat. no.:76498-140
Spécifications
A propos de cet article
This user-friendly, portable instrument offers high resolution and accurate precision measurements for pH, mV, conductivity, and oxygen in on-site applications. It can measure a maximum of 2 of these parameters simultaneously.
- Large graphic LCD with continuous LED backlight showing both pH/mV and temperature value simultaneously
- Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets; additional 200 memory points with manual storage
- Automatic power shut-off: Adjustable from 10 min to 24 h
- Automatic buffer recognition of DIN and NIST buffers (1.68/4.00/6.86/9.18/12.54)
- Three additional technical buffers at 25 °C (4.00/7.00/10.00) in memory
- Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
- Low energy consumption: Choice of mains supply or 4× AA batteries for over 1000 operating hours
With features that include waterproof USB output for connection to a computer and a rugged IP 67 enclosure.
The MU 6100 H S2 kit (76498-142) also includes the OXY 11-3 galvanic oxygen sensor with an integrated temperature sensor (3 m cable) and a maintenance kit.
76460-522 includes meter, case, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual.