ContactUsLinkComponent
Flow Cell(path: 5mm, vol.: 3.2ul, pressure:1.0MPa), semi micro, For 5410/5420
# de catalogue 76493-152
Flow Cell(path: 5mm, vol.: 3.2ul, pressure:1.0MPa), semi micro, For 5410/5420
# de catalogue 76493-152
Customers Who Bought This Also Bought