Order Entry
ContactUsLinkComponent
VWR Electronic Timer, 4-Channel With White Board And Certificate Of Calibration
VWR® 4-Channel Switchable Electronic Timer
# de catalogue 76491-692
Fournisseur:  VWR International
VWR® 4-Channel Switchable Electronic Timer
# de catalogue 76491-692
Fournisseur:  VWR International

Spécifications

  • Description:
    Digital timer, 4 channels, with white board
  • Cat. no.:
    76491-692
  • W×H×D:
    114.3×25.4×88.9 mm
  • Accuracy:
    ±5 s/day

Spécifications

A propos de cet article

Timer features a built-in a whiteboard with marker and eraser, which helps to keep track of time and other critical information.

  • Switchable count up and down with elapsed time displayed after countdown
  • Switchable capacity of 23 hours, 59 minutes to 59 minutes, 59 seconds
  • Audible alarm for each channel
  • Last time set memory
  • Audible confirmation for a data entry key press

Mounting options: lay flat on bench, flip-open stand for vertical view, and a magnetic back for metal surface attachment.

Certifications: Individually tested and inspected using NIST in A2LA accredited laboratory (certification #2448.01), assuring conformance to ISO 17025:2005. RoHS compliant.
Delivery: Supplied with batteries and certificate of calibration. Requires two AAA batteries.
Customers Who Bought This Also Bought