VWR® Environmentally Friendly Thermometers, Double Safe
# de catalogue 76491-608
Fournisseur: VWR International
Spécifications
- Accuracy:±1.0, 1.5 (>100 °C)
- Length:200 mm
- Cat. no.:76491-608
- Divisions:1.0 °C
- Range:−10...+150 °C
- Immersion:50 mm
Spécifications
A propos de cet article
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. The numbers are printed larger than on a standard thermometer. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
- Enviro safe liquid
- Non-toxic, non-hazardous, biodegradable liquid
- PFA coating prevents any contamination of liquid or glass
Includes a multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability.
Certifications: Traceable to NIST.
Customers Who Bought This Also Bought