VWR® Environmentally Friendly Thermometers, Easy Read
# de catalogue 76491-598
Fournisseur: VWR International
Spécifications
- Accuracy:±1.0 °C
- Length:305 mm
- Cat. no.:76491-598
- Divisions:1.0 °C
- Range:−20...+110 °C
- Immersion:76 mm
Spécifications
A propos de cet article
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
- Enviro safe liquid
- Eliminates reading errors
- Non-toxic, non-hazardous, biodegradable liquid
Includes multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability, non roll fitting and individual serial number.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).
Customers Who Bought This Also Bought