Order Entry
ContactUsLinkComponent
Tube, Test, Reusable, Plain End, 170mL, 38mm x 200mm (OD x H) ISO/DIN 4142, round Bottom, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 170mL, Approx O.D. X Length: 38mm x 200mm, Class/ Quality Grade: Type I,
Borosil® Reusable Test Tubes without Rim, Foxx Life Sciences
# de catalogue 76491-546
Fournisseur:  Foxx Life Sciences
Tube, Test, Reusable, Plain End, 170mL, 38mm x 200mm (OD x H) ISO/DIN 4142, round Bottom, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 170mL, Approx O.D. X Length: 38mm x 200mm, Class/ Quality Grade: Type I,
Borosil® Reusable Test Tubes without Rim, Foxx Life Sciences
# de catalogue 76491-546
Fournisseur:  Foxx Life Sciences

Spécifications

  • Volume:
    170 ml
  • Description:
    Borosil® reusable test tubes with plain end
  • Cat. no.:
    76491-546
  • O.D.×L:
    38×200 mm

Spécifications

A propos de cet article

Test tubes constructed of borosilicate 3.3 glass, without rim.

  • Excellent chemical resistance
  • Thick walled, mechanically resistant
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Designed without a rim
  • Graduation range: Type I

The large opening can even accommodate large volume funnels, giving easy access to scoops or spatulas.

Amber to protect light-sensitive materials.

Certifications: In compliance with ISO/DIN 4142 standards.
Customers Who Bought This Also Bought