ContactUsLinkComponent
Amber Culture Tube Media Flat Bottom PP cap with Liner 15 mL icate CS/100, Material: 3.3 Borosilicate Glass, Tube size: 15 mL, Color: Amber, Cap Material: Polypropylene, Overall Dimension: 9911007, Class/Quality Grade: Type 1, Class A , Documentation, Datasheets Coming Soon!
# de catalogue 76491-022
Fournisseur:  Foxx Life Sciences
Amber Culture Tube Media Flat Bottom PP cap with Liner 15 mL icate CS/100, Material: 3.3 Borosilicate Glass, Tube size: 15 mL, Color: Amber, Cap Material: Polypropylene, Overall Dimension: 9911007, Class/Quality Grade: Type 1, Class A , Documentation, Datasheets Coming Soon!
Amber Culture Tube Media Flat Bottom PP cap with Liner 15 mL icate CS/100, Material: 3.3 Borosilicate Glass, Tube size: 15 mL, Color: Amber, Cap Material: Polypropylene, Overall Dimension: 9911007, Class/Quality Grade: Type 1, Class A , Documentation, Datasheets Coming Soon!
# de catalogue 76491-022
Fournisseur:  Foxx Life Sciences
Customers Who Bought This Also Bought