ContactUsLinkComponent
Media Amber Culture Tube, Flat Bottom, PP Screw Cap with PTFE Liner, 5 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Amber, Capacity: 5mL, Approx O.D. X Length: 16mm x 50mm, Class/ Quality Grade: Type I, Class A,
# de catalogue 76491-020
Fournisseur:  Foxx Life Sciences
Media Amber Culture Tube, Flat Bottom, PP Screw Cap with PTFE Liner, 5 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Amber, Capacity: 5mL, Approx O.D. X Length: 16mm x 50mm, Class/ Quality Grade: Type I, Class A,
Media Amber Culture Tube, Flat Bottom, PP Screw Cap with PTFE Liner, 5 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Amber, Capacity: 5mL, Approx O.D. X Length: 16mm x 50mm, Class/ Quality Grade: Type I, Class A,
# de catalogue 76491-020
Fournisseur:  Foxx Life Sciences
Customers Who Bought This Also Bought