Amber Culture Tube Media Round Bottom PP cap with Liner 60 mL icate CS/50, Material: 3.3 Borosilicate Glass, Tube size: 60 mL, Color: Amber, Cap Material: Polypropylene, Overall Dimension: 9901013, Class/Quality Grade: Type I, Class A, Documentation, Datasheets Coming Soon!
# de catalogue 76491-008
Fournisseur: Foxx Life Sciences
Customers Who Bought This Also Bought