ContactUsLinkComponent
Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 2L, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 2L, Approx O.D. X Height: 168mm x 270mm, Class/ Quality Grade: Type I, neck finish is developed for strength and to ensure a proper stopper fitting
# de catalogue 76490-808
Fournisseur:  Foxx Life Sciences
Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 2L, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 2L, Approx O.D. X Height: 168mm x 270mm, Class/ Quality Grade: Type I, neck finish is developed for strength and to ensure a proper stopper fitting
Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 2L, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 2L, Approx O.D. X Height: 168mm x 270mm, Class/ Quality Grade: Type I, neck finish is developed for strength and to ensure a proper stopper fitting
# de catalogue 76490-808
Fournisseur:  Foxx Life Sciences
Customers Who Bought This Also Bought