ContactUsLinkComponent
Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 1L, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 1L, Approx O.D. X Height: 136mm x 227mm, Class/ Quality Grade: Type I,
# de catalogue 76490-806
Fournisseur:  Foxx Life Sciences
Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 1L, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 1L, Approx O.D. X Height: 136mm x 227mm, Class/ Quality Grade: Type I,
Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 1L, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 1L, Approx O.D. X Height: 136mm x 227mm, Class/ Quality Grade: Type I,
# de catalogue 76490-806
Fournisseur:  Foxx Life Sciences
Customers Who Bought This Also Bought