Vibration Dampening Mounts, METTLER TOLEDO®
# de catalogue 76492-554
Fournisseur: Mettler Toledo
Spécifications
- Description:Marble slab, polished
- Height:2"
- Cat. no.:76492-554
- W×D:20×13"
- Weight:27.2 kg (60 lbs.)
Spécifications
A propos de cet article
Vibrations can negatively influence weighing results. A proper balance table or marble slab is necessary to ensure ergonomic and efficient weighing in a lab.
- Speeds up the working process
- Increases the quality of results
Anti-vibration tables enable proper balance operation and faster stabilization times.
Customers Who Bought This Also Bought