Borosil® Culture/Petri Dishes, with Cover, Borosilicate Glass 3.3, Foxx Life Sciences
# de catalogue 76490-724
Fournisseur: Foxx Life Sciences
Spécifications
- Description:Borosil® Culture/Petri dish, with cover
- Material:Borosilicate glass 3.3
- Cat. no.:76490-724
- O.D.×H:150×20 mm
Spécifications
A propos de cet article
Borosil® Petri dishes have consistent wall thickness for optical performance.
- Material: Borosilicate glass 3.3
- Color: Clear
- Class/Quality grade: Type I
Borosil® brand Dishes are designed to fulfill wide-ranging needs from crystalizing, evaporating, drying, and storing to Petri dishes for culture, assay and micro-biological works. These are ideal for use in routine work. These Dishes are clear and withstand repeat sterilization. Each is formed to ensure even wall thickness and consistent optical performance. Borosil® brand Dishes are safe, durable and economical.
Certifications: Comply with IS 2626.
Customers Who Bought This Also Bought