MXP Data Loggers, Cryopak Verification Technologies
# de catalogue 76490-420
Fournisseur: Cryopak Verification Technologies
Spécifications
- Description:Data logger, MXP single use
- Cat. no.:76490-420
Spécifications
A propos de cet article
The MXP data logger offers flexibility and longevity with a light sensor to detect and log if package is opened.
- Long shelf life
- Event logging feature
- Internal digital temperature sensor
With its USB connectivity, the MXP data logger provides detailed reports in both PDF and Excel format, and is user friendly for both Windows and smartphone app use.
Customers Who Bought This Also Bought