Order Entry
ContactUsLinkComponent
Gown, Non-sterile, level 2 SMS, moderate fluid resistant, breathable, knee length, tie at back, knit cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, Medical Blue, Reg size fits to XL
AAMI Level 2 SMS Gowns, Apex Aseptic Products
# de catalogue 76488-400
Gown, Non-sterile, level 2 SMS, moderate fluid resistant, breathable, knee length, tie at back, knit cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, Medical Blue, Reg size fits to XL
AAMI Level 2 SMS Gowns, Apex Aseptic Products
# de catalogue 76488-400

Spécifications

  • Clothing Size:
    Regular
  • Color:
    Medical Blue
  • Cat. no.:
    76488-400

Spécifications

A propos de cet article

Fluid resistant, breathable, and comfortable SMS Isolation Gowns meet AAMI Level 2 Standards.

  • Hydrohead test result: 350 mm
  • Spray Impact Rating: 0.8 g
  • Latex-free, TSE-free, BSE-free
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Manufactured in controlled environment

The entire gown is made with Level 2 SMS fabrics, ensuring the garment meets AAMI Level 2 Hydrohead and Spray Impact standards. These knee length gowns feature a full back, knit cuffs, and back ties that allow for adjustable sizing.

Certifications: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
Packaging: Gowns are packaged 10 per pack, 100 per case.
Customers Who Bought This Also Bought