Order Entry
ContactUsLinkComponent
Packer Bottle, Clear PET with 45-400 White Polypropylene SturdeeSeal® PE Foam Lined Cap attached, Vacuum & Ionized, 17oz (500ml
Packer Bottles, PET, Clear, Wide Mouth, Qorpak
# de catalogue 76487-362
Fournisseur:  Qorpak
Packer Bottle, Clear PET with 45-400 White Polypropylene SturdeeSeal® PE Foam Lined Cap attached, Vacuum & Ionized, 17oz (500ml
Packer Bottles, PET, Clear, Wide Mouth, Qorpak
# de catalogue 76487-362
Fournisseur:  Qorpak

Spécifications

  • Cap Size:
    45-400
  • Cap Type:
    White polypropylene F217 lined
  • Capacity:
    500 ml (16.9 oz.)
  • Color:
    Clear
  • Description:
    Vacuum and ionized
  • Material:
    PET
  • Cat. no.:
    76487-362

Spécifications

A propos de cet article

PET Packer Bottles feature glass like clarity and are durable and rigid.

  • PET packers are stress and impact resistant
  • PET bottles are ideal for oil analysis

They have exceptional gas and moisture barrier properties and a good alcohol and solvent barrier.

Ordering information: Bottle 30617-180 is Vacuum and Ionized. Bottle 89234-738 is Super Cleaned for Oil Analysis.
Customers Who Bought This Also Bought