ContactUsLinkComponent
Pipette Tip, Robotic Sterile Filtered; Hamilton Type, 1000uL, Conductive, Carbon filled Polypropylene, For automated liquid handling equipment
SP Bel-Art Robotic Sterile Pipette Tips, Bel-Art Products, a part of SP
Catalog # 76487-094
Pipette Tip, Robotic Sterile Filtered; Hamilton Type, 1000uL, Conductive, Carbon filled Polypropylene, For automated liquid handling equipment
SP Bel-Art Robotic Sterile Pipette Tips, Bel-Art Products, a part of SP
Catalog # 76487-094
Specifications
  • Volume:
    1000 µl
  • Tip Color:
    Black
  • Packed:
    16 Racks of 96 Tips
  • Cat. no.:
    76487-094
  • Tip Style:
    Conductive, carbon filled PP
  • For Use With:
    Hamilton® STAR™, STARlet, and NIMBUS®
Specifications
About this item
Designed for use with automated liquid handling equipment, these sterile, disposable, filtered pipette tips can be used in robotic systems as indicated.
  • DNase, RNase, pyrogen free; manufactured in Class 8 clean room
  • Filtered to prevent contamination of pipette arm
Conductive tip models detect the liquid level when pipetting conductive liquids to ensure repeatable volume accuracy.
Customers Who Bought This Also Bought