SP Bel-Art Robotic Sterile Pipette Tips, Bel-Art Products, a part of SP
Catalog # 76487-094
Supplier: Bel-Art Products, a Part of SP
Specifications
- Volume:1000 µl
- Tip Color:Black
- Packed:16 Racks of 96 Tips
- Cat. no.:76487-094
- Tip Style:Conductive, carbon filled PP
- For Use With:Hamilton® STAR™, STARlet, and NIMBUS®
Specifications
About this item
Designed for use with automated liquid handling equipment, these sterile, disposable, filtered pipette tips can be used in robotic systems as indicated.
- DNase, RNase, pyrogen free; manufactured in Class 8 clean room
- Filtered to prevent contamination of pipette arm
Conductive tip models detect the liquid level when pipetting conductive liquids to ensure repeatable volume accuracy.
Customers Who Bought This Also Bought