Boot Cover, SBP White, Standard Weight, Spunbond Polypropylene shoe covers offers lightweight protection against dry particulates, Roomy construction to fit most boot sizes, Used to keep dirt off or on the shoe, Elastic band at top of boot, 18in length
# de catalogue 76486-356
Fournisseur: PIP
Customers Who Bought This Also Bought