Disposable Transfer Pipettes
# de catalogue 470334-420
Fournisseur: GSC International
Spécifications
- Length:300 mm
- Volume:23 ml
- Description:Disposable transfer pipette
Spécifications
A propos de cet article
Used in many applications such as urinalysis, wet chemistry, microbiology, hematology, and serology.
- Small micro-tip
- Made from LDPE
Ensure a clean, contamination-free experience every time. Whether working in a lab, a medical facility, or an industrial setting, this is an essential tool for STEM professionals.
Customers Who Bought This Also Bought