ContactUsLinkComponent
VWR M-Series Moisture Analyzer Aluminum Sample Pans, 90mm
VWR® Disposable Aluminum Weighing and Drying Pans
# de catalogue 76485-880
Fournisseur:  VWR International
VWR M-Series Moisture Analyzer Aluminum Sample Pans, 90mm
VWR® Disposable Aluminum Weighing and Drying Pans
# de catalogue 76485-880
Fournisseur:  VWR International
Spécifications
  • Description:
    Weighing pans, aluminum, Ø 90 mm
  • Cat. no.:
    76485-880