FreezeBox CF 5mL System, Blue, Package includes 1 each of FZB-S1 FreezeBox, CF Freeze Core -18C to -4C, and CM-03 Tube Module for 30 x 5ml cryo tubes, 152 x 170 x 123mm, Uniform, low temperature protection for consistent sample results, typically +/-0.1 C
# de catalogue 76480-462
Fournisseur: CRYSTAL TECHNOLOGY & INDUSTRIES
Customers Who Bought This Also Bought