ContactUsLinkComponent
VWR Tubing, 70A, Material: PVC, 1.5X2 50Ft, phthalate, DEHP, BPA free. It ensures the safe transfer of fluids, gases, air in the food, beverage, medical device, health care markets, NSF-51 listed, meets USP Class VI requirements, is made from non-toxic ingredients conforming to FDA, 3A standards.
# de catalogue 76480-182
Fournisseur:  VWR International
VWR Tubing, 70A, Material: PVC, 1.5X2 50Ft, phthalate, DEHP, BPA free. It ensures the safe transfer of fluids, gases, air in the food, beverage, medical device, health care markets, NSF-51 listed, meets USP Class VI requirements, is made from non-toxic ingredients conforming to FDA, 3A standards.
# de catalogue 76480-182
Fournisseur:  VWR International
Customers Who Bought This Also Bought