VWR Tubing, 70A, Material: PVC, 1/2X5/8 100Ft, phthalate, DEHP, BPA free. It ensures safe transfer of fluids, gases, air in food, beverage, medical device, health care markets, NSF-51 listed, meets USP Class VI requirements, is made from non-toxic ingredients conforming to FDA, 3A standards.
# de catalogue 76480-150
Fournisseur: VWR International
Customers Who Bought This Also Bought