Humidity control, with Dehum, from ambient to 85% RH for 7300 & 7310 22 ft3 series. Includes humidity display/readout. Actively humidifies with gVapor and dehumidifies with evaporator. Factory installed at the time of order.
# de catalogue 76479-106
Fournisseur: Caron Products
Customers Who Bought This Also Bought