Extended temperature, range down to +5C (lights on) for R290 7540/7545 chambers. Factory installed at time of order.
# de catalogue 76479-104
Fournisseur: Caron Products
Extended temperature, range down to +5C (lights on) for R290 7540/7545 chambers. Factory installed at time of order.
# de catalogue 76479-104
Fournisseur: Caron Products
Customers Who Bought This Also Bought
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Fournisseur: VWR International
$89.92 to $1,797.48
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$639.34 to $1,284.30