Dust/Dry Mop, Green with White Trim, 18in, provides long-lasting, dependable cleaning power, made of positively charged fibers that attract and trap negatively charged dirt particles, Microfiber combined with high density foam inner layer for increased stability
# de catalogue 76478-818
Fournisseur: Medline
Customers Who Bought This Also Bought