Cole-Parmer® Essentials Frequent-Use Hand Soap, Cole-Parmer
# de catalogue 76478-602
Fournisseur: Argos Technologies
Spécifications
- Capacity:500 ml (17 oz.)
- Description:Cole-Parmer® Essentials frequent-use hand soap, unscented, pump dispenser
- Cat. no.:76478-602
Spécifications
A propos de cet article
Reduce cross-contamination with this laboratory grade soap.
- Gently and effectively washes away the most common germs for better hand health
- Contains natural moisturizers like aloe and lanolin to nourish skin
The soothing coconut-based formula with moisturizing lanolin and aloe is gentle on skin, so it cleans without drying or chapping. Used by scientists who wash their hands up to 40 times a day. Available in a 17 oz. (500 ml) convenient pump dispenser.
Customers Who Bought This Also Bought