ContactUsLinkComponent
LED FLASHLIGHT
Flashlight, Led, Assorted Colors, United Scientific
# de catalogue 470332-808
LED FLASHLIGHT
Flashlight, Led, Assorted Colors, United Scientific
# de catalogue 470332-808
Spécifications
  • Description:
    LED flashlight, assorted colors
  • Cat. no.:
    470332-808
Spécifications
A propos de cet article
Plastic 6" long flashlight includes long-lasting LED illumination and lanyard.
  • Uses two AA batteries (not included)
  • Assorted colors
This flashlight allows illumination experiments with various colors. The LEDs insure a long life span and the lanyard makes it easy to hang from a support rod.
Customers Who Bought This Also Bought