Bottles With Dropper, Square, Flint Glass, United Scientific Supplies
# de catalogue 470332-792
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:1 oz.
- Color:Clear
- Description:Bottles with dropper, square, flint glass
- Material:Flint glass
- Cat. no.:470332-792
Spécifications
A propos de cet article
Pack of 12 square glass dropping bottles.
- Non-porous glass is easy to clean and autoclavable
These square flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving.
Ordering information: Screw cap with dropper assembly included.
Customers Who Bought This Also Bought