ContactUsLinkComponent
BOTTLES W/ DROPPER, BOSTON, AMBER, 0.5OZ
Bottles with Dropper, Boston Round, Amber, United Scientific Supplies
Catalog # 470332-786
BOTTLES W/ DROPPER, BOSTON, AMBER, 0.5OZ
Bottles with Dropper, Boston Round, Amber, United Scientific Supplies
Catalog # 470332-786
Specifications
  • Capacity:
    0.5 oz.
  • Color:
    Amber
  • Description:
    Bottles with dropper, Boston round
  • Cat. no.:
    470332-786
Specifications
About this item
Amber Boston round bottle with droppers for general use.
  • Screw cap with dropper assembly included
  • Non-porous glass is easy to clean and autoclavable
  • Amber color protects light-sensitive contents
These amber Boston round flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving. The amber color helps protect light sensitive materials.
Customers Who Bought This Also Bought