ContactUsLinkComponent
Scrubbing Sponge, Light duty, White, feature a cellulose sponge on one side and a scouring pad on the other, Cellulose sponge absorbs water while the pad helps clean, Measure 6.25in x 3.18in x 0.88in
# de catalogue 76478-748
Fournisseur:  Medline
Scrubbing Sponge, Light duty, White, feature a cellulose sponge on one side and a scouring pad on the other, Cellulose sponge absorbs water while the pad helps clean, Measure 6.25in x 3.18in x 0.88in
# de catalogue 76478-748
Fournisseur:  Medline
Customers Who Bought This Also Bought