ContactUsLinkComponent
Floor Pad, Burnishing, UHS, Ice Blue, 27in, Ideal for top dressing highly polished floors, Recommended for use on machines up to 3,000 RPM
# de catalogue 76478-722
Fournisseur:  Medline
Floor Pad, Burnishing, UHS, Ice Blue, 27in, Ideal for top dressing highly polished floors, Recommended for use on machines up to 3,000 RPM
# de catalogue 76478-722
Fournisseur:  Medline
Customers Who Bought This Also Bought