Floor Polishing Pad, Extra-Fine, Ultra-High-Speed, for polishing clean, dry floors, Use to buff soft finishes and low traffic areas, Ideal for marble floors, Machine speed: up to 3,000 RPM, White, 20in
# de catalogue 76478-714
Fournisseur: Medline
Customers Who Bought This Also Bought